Order Entry
United Kingdom
ContactUsLinkComponent
GCN2-IN-1 (A-92) is a potent general control nonderepressible 2 kinase (GCN2) inhibitor with an IC50 of IC50 of 0.3-3 μM in the cell assay[1]. 1 * 5 mg
Catalog # MDEXHY-100877
Supplier:  MEDCHEMEXPRESS
GCN2-IN-1 (A-92) is a potent general control nonderepressible 2 kinase (GCN2) inhibitor with an IC50 of IC50 of 0.3-3 μM in the cell assay[1]. 1 * 5 mg
Catalog # MDEXHY-100877
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together