Order Entry
United Kingdom
ContactUsLinkComponent
Denifanstat (TVB-2640) is an orally active and potent Fatty Acid Synthase (FASN) inhibitor with an IC50 of 0.052 μM and an EC50 of 0 .072 μM. Denifanstat has the potential for fatty liver disease and cancer research[1][2]. 1 * 5 mg
Catalog # MDEXHY-112829
Supplier:  MEDCHEMEXPRESS
Denifanstat (TVB-2640) is an orally active and potent Fatty Acid Synthase (FASN) inhibitor with an IC50 of 0.052 μM and an EC50 of 0 .072 μM. Denifanstat has the potential for fatty liver disease and cancer research[1][2]. 1 * 5 mg
Catalog # MDEXHY-112829
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together