Order Entry
United Kingdom
ContactUsLinkComponent
AR-C155858 is a selective monocarboxylate transporter MCT1 and MCT2 inhibitor with Kis of 2.3 nM and 10 nM, respectively. 1 * 100 mg
Catalog # MDEXHY-13248
Supplier:  MEDCHEMEXPRESS
AR-C155858 is a selective monocarboxylate transporter MCT1 and MCT2 inhibitor with Kis of 2.3 nM and 10 nM, respectively. 1 * 100 mg
Catalog # MDEXHY-13248
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together