Order Entry
United Kingdom
ContactUsLinkComponent
Navtemadlin (AMG 232) is a potent, selective and orally available inhibitor of p53-MDM2 interaction, with an IC50 of 0.6 nM. Navtema dlin binds to MDM2 with a Kd of 0.045 nM[1][2]. 1 * 100 mg
Catalog # MDEXHY-12296
Supplier:  MEDCHEMEXPRESS
Navtemadlin (AMG 232) is a potent, selective and orally available inhibitor of p53-MDM2 interaction, with an IC50 of 0.6 nM. Navtema dlin binds to MDM2 with a Kd of 0.045 nM[1][2]. 1 * 100 mg
Catalog # MDEXHY-12296
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together