Order Entry
United Kingdom
ContactUsLinkComponent
Pirfenidone (AMR69) is an antifibrotic agent that attenuates CCL2 and CCL12 production in fibrocyte cells. Pirfenidone has growth-in hibitory effect and reduces TGF-β2 protein levels in human glioma cell lines. Pirfenidone also has anti-inflammatory 1 * 5 g
Catalog # MDEXHY-B0673
Supplier:  MEDCHEMEXPRESS
Pirfenidone (AMR69) is an antifibrotic agent that attenuates CCL2 and CCL12 production in fibrocyte cells. Pirfenidone has growth-in hibitory effect and reduces TGF-β2 protein levels in human glioma cell lines. Pirfenidone also has anti-inflammatory 1 * 5 g
Catalog # MDEXHY-B0673
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together