Order Entry
United Kingdom
ContactUsLinkComponent
TDI-011536 is a potent Lats kinase inhibitor, interrupts Hippo-Yap signaling and initiates the proliferation of lesioned heartmuscle cells. TDI-011536 can be used in studies of organ conservation and regeneration[1]. 1 * 25 mg
  MDEXHY-150042
TDI-011536 is a potent Lats kinase inhibitor, interrupts Hippo-Yap signaling and initiates the proliferation of lesioned heartmuscle cells. TDI-011536 can be used in studies of organ conservation and regeneration[1]. 1 * 25 mg
  MDEXHY-150042
Frequently Bought Together