Order Entry
United Kingdom
ContactUsLinkComponent
TJ-M2010-5 is a MyD88 inhibitor that binds to the TIR domain of MyD88 to interfere with its homodimerization, and the TLR/MyD88 sign al pathway[1][2]. TJ-M2010-5 can be used for the research of myocardial ischemia/reperfusion injury (MIRI)[2]. 1 * 10 mg
Catalog # MDEXHY-139397
Supplier:  MEDCHEMEXPRESS
TJ-M2010-5 is a MyD88 inhibitor that binds to the TIR domain of MyD88 to interfere with its homodimerization, and the TLR/MyD88 sign al pathway[1][2]. TJ-M2010-5 can be used for the research of myocardial ischemia/reperfusion injury (MIRI)[2]. 1 * 10 mg
Catalog # MDEXHY-139397
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together