Order Entry
United Kingdom
ContactUsLinkComponent
Ertapenem sodium (L-749345) is a broad spectrum and long acting β-lactam antibiotic. Ertapenem sodium has a broad-spectrum anti-anae robic activity against a variety of anaerobes with a mode MIC of 0.12 μg/mL. Ertapenem sodium can be used for the res 1 * 25 mg
Catalog # MDEXHY-13625
Supplier:  MEDCHEMEXPRESS
Ertapenem sodium (L-749345) is a broad spectrum and long acting β-lactam antibiotic. Ertapenem sodium has a broad-spectrum anti-anae robic activity against a variety of anaerobes with a mode MIC of 0.12 μg/mL. Ertapenem sodium can be used for the res 1 * 25 mg
Catalog # MDEXHY-13625
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together