Order Entry
United Kingdom
ContactUsLinkComponent
STC-15 is an orally active RNA methyltransferase METTL3 inhibitor with the activity of activating anti-tumor immunity and reshaping the tumor microenvironment. STC-15 inhibits tumor growth by activating anti-cancer immune responses associated with in 1 * 5 mg
Catalog # MDEXHY-156677
Supplier:  MEDCHEMEXPRESS
STC-15 is an orally active RNA methyltransferase METTL3 inhibitor with the activity of activating anti-tumor immunity and reshaping the tumor microenvironment. STC-15 inhibits tumor growth by activating anti-cancer immune responses associated with in 1 * 5 mg
Catalog # MDEXHY-156677
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together