Order Entry
United Kingdom
ContactUsLinkComponent
IBMX is a broad-spectrum phosphodiesterase (PDE) inhibitor, with IC50s of 6.5, 26.3 and 31.7 μM for PDE3, PDE4 and PDE5, respectivel y. 1 * 200 mg
Catalog # MDEXHY-12318
Supplier:  MEDCHEMEXPRESS
IBMX is a broad-spectrum phosphodiesterase (PDE) inhibitor, with IC50s of 6.5, 26.3 and 31.7 μM for PDE3, PDE4 and PDE5, respectivel y. 1 * 200 mg
Catalog # MDEXHY-12318
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together