Order Entry
United Kingdom
ContactUsLinkComponent
EED226 is a polycomb repressive complex 2 (PRC2) inhibitor, which binds to the K27me3-pocket on embryonic ectoderm development (EED) and shows strong antitumor activity in xenograft mice model[1]. EED226 is a potent, selective, and orally bioavailab 1 * 25 mg
Catalog # MDEXHY-101117
Supplier:  MEDCHEMEXPRESS
EED226 is a polycomb repressive complex 2 (PRC2) inhibitor, which binds to the K27me3-pocket on embryonic ectoderm development (EED) and shows strong antitumor activity in xenograft mice model[1]. EED226 is a potent, selective, and orally bioavailab 1 * 25 mg
Catalog # MDEXHY-101117
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together