Order Entry
United Kingdom
ContactUsLinkComponent
Iberdomide (CC-220) is an orally active and potent cereblon (CRBN) E3 ligase modulator (CELMoD) with an IC50 of ~150 nM for cereblon -binding affinity. Iberdomide, a derivative of Thalidomide (HY-14658), has antitumor and immunostimulatory activities 1 * 50 mg
Catalog # MDEXHY-101291
Supplier:  MEDCHEMEXPRESS
Iberdomide (CC-220) is an orally active and potent cereblon (CRBN) E3 ligase modulator (CELMoD) with an IC50 of ~150 nM for cereblon -binding affinity. Iberdomide, a derivative of Thalidomide (HY-14658), has antitumor and immunostimulatory activities 1 * 50 mg
Catalog # MDEXHY-101291
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together