Order Entry
United Kingdom
ContactUsLinkComponent
PD98059 is a potent and selective MEK inhibitor with an IC50 of 5 µM. PD98059 binds to the inactive form of MEK, thereby preventing the activation of MEK1 (IC50 of 2-7 µM) and MEK2 (IC50 of 50 µM) by upstream kinases. PD98059 is a ERK1/2 signaling in 1 * 100 mg
Catalog # MDEXHY-12028
Supplier:  MEDCHEMEXPRESS
PD98059 is a potent and selective MEK inhibitor with an IC50 of 5 µM. PD98059 binds to the inactive form of MEK, thereby preventing the activation of MEK1 (IC50 of 2-7 µM) and MEK2 (IC50 of 50 µM) by upstream kinases. PD98059 is a ERK1/2 signaling in 1 * 100 mg
Catalog # MDEXHY-12028
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together