Order Entry
United Kingdom
ContactUsLinkComponent
Pazopanib (GW786034) is a novel multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRβ, c-Kit, FGFR1, and c-Fms with IC50s of 10, 30, 47, 84, 74, 140 and 146 nM, respectively. 1 * 500 mg
Catalog # MDEXHY-10208
Supplier:  MEDCHEMEXPRESS
Pazopanib (GW786034) is a novel multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRβ, c-Kit, FGFR1, and c-Fms with IC50s of 10, 30, 47, 84, 74, 140 and 146 nM, respectively. 1 * 500 mg
Catalog # MDEXHY-10208
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together