Order Entry
United Kingdom
ContactUsLinkComponent
Flavopiridol (Alvocidib) is a broad spectrum and competitive inhibitor of CDKs, inhibiting CDK1, CDK2, CDK4 with IC50s of 30, 170, 1 00 nM, respectively. 1 * 100 mg
  MDEXHY-10005
Flavopiridol (Alvocidib) is a broad spectrum and competitive inhibitor of CDKs, inhibiting CDK1, CDK2, CDK4 with IC50s of 30, 170, 1 00 nM, respectively. 1 * 100 mg
  MDEXHY-10005
Frequently Bought Together