Order Entry
United Kingdom
ContactUsLinkComponent
Roxadustat is an orally active hypoxia-inducible factor (HIF) prolyl-hydroxylase (PHD) inhibitor (HIF-PHI) that promotes erythropoie sis through increasing endogenous erythropoietin, improving iron regulation, and reducing hepcidin[1]. 1 * 100 mg
  MDEXHY-13426
Roxadustat is an orally active hypoxia-inducible factor (HIF) prolyl-hydroxylase (PHD) inhibitor (HIF-PHI) that promotes erythropoie sis through increasing endogenous erythropoietin, improving iron regulation, and reducing hepcidin[1]. 1 * 100 mg
  MDEXHY-13426
Frequently Bought Together