Order Entry
United Kingdom
ContactUsLinkComponent
Brigatinib (AP-26113) is a highly potent, selective and orally active ALK inhibitor, with an IC50 of 0.6 nM. Brigatinib can be used for research of NSCLC[1]. 1 * 10 mg
Catalog # MDEXHY-12857
Supplier:  MEDCHEMEXPRESS
Brigatinib (AP-26113) is a highly potent, selective and orally active ALK inhibitor, with an IC50 of 0.6 nM. Brigatinib can be used for research of NSCLC[1]. 1 * 10 mg
Catalog # MDEXHY-12857
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together