Order Entry
United Kingdom
ContactUsLinkComponent
Entrectinib (NMS-E628) is an orally active, BBB-penetrated and centrally active inhibitor of TrkA/B/C, ROS1 and ALK, with IC50 value s of 1, 3, 5, 12 and 7 nM, respectively. Entrectinib induces apoptosis and cycle arrest in cancer cells, has antitumo 1 * 100 mg
Catalog # MDEXHY-12678
Supplier:  MEDCHEMEXPRESS
Entrectinib (NMS-E628) is an orally active, BBB-penetrated and centrally active inhibitor of TrkA/B/C, ROS1 and ALK, with IC50 value s of 1, 3, 5, 12 and 7 nM, respectively. Entrectinib induces apoptosis and cycle arrest in cancer cells, has antitumo 1 * 100 mg
Catalog # MDEXHY-12678
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together