Order Entry
United Kingdom
ContactUsLinkComponent
Sotuletinib (BLZ945) is a potent, selective and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM, showing more than 1,0 00-fold selectivity against its closest receptor tyrosine kinase homologs[1]. 1 * 100 mg
Catalog # MDEXHY-12768
Supplier:  MEDCHEMEXPRESS
Sotuletinib (BLZ945) is a potent, selective and brain-penetrant CSF-1R (c-Fms) inhibitor with an IC50 of 1 nM, showing more than 1,0 00-fold selectivity against its closest receptor tyrosine kinase homologs[1]. 1 * 100 mg
Catalog # MDEXHY-12768
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together