Order Entry
United Kingdom
ContactUsLinkComponent
Palbociclib (PD 0332991) monohydrochloride is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, r espectively. Palbociclib monohydrochloride has potent anti-proliferative activity and induces cell cycle arrest in ca 1 * 200 mg
Catalog # MDEXHY-50767A
Supplier:  MEDCHEMEXPRESS
Palbociclib (PD 0332991) monohydrochloride is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, r espectively. Palbociclib monohydrochloride has potent anti-proliferative activity and induces cell cycle arrest in ca 1 * 200 mg
Catalog # MDEXHY-50767A
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together