Order Entry
United Kingdom
ContactUsLinkComponent
CC-885 is a cereblon (CRBN) modulator with potent anti-tumour activity. CC-885 is also a known degrader of GSPT1, inhibiting protein translation. 1 * 10 mg
  MDEXHY-101488
CC-885 is a cereblon (CRBN) modulator with potent anti-tumour activity. CC-885 is also a known degrader of GSPT1, inhibiting protein translation. 1 * 10 mg
  MDEXHY-101488
Frequently Bought Together