Order Entry
United Kingdom
ContactUsLinkComponent
Elimusertib (BAY-1895344) is a potent, orally active and selective ATR inhibitor with an IC50 of 7 nM. Elimusertib has anti-tumor ac tivity[1][2]. Elimusertib can be used for the research of solid tumors and lymphomas[3]. 1 * 50 mg
Catalog # MDEXHY-101566
Supplier:  MEDCHEMEXPRESS
Elimusertib (BAY-1895344) is a potent, orally active and selective ATR inhibitor with an IC50 of 7 nM. Elimusertib has anti-tumor ac tivity[1][2]. Elimusertib can be used for the research of solid tumors and lymphomas[3]. 1 * 50 mg
Catalog # MDEXHY-101566
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together