Order Entry
United Kingdom
ContactUsLinkComponent
Mezigdomide (CC-92480), a cereblon E3 ubiquitin ligase modulating agent (CELMoD), acts as a molecular glue. Mezigdomide shows high a ffinity to cereblon, resulting in potent antimyeloma activity[1]. 1 * 25 mg
  MDEXHY-129395
Mezigdomide (CC-92480), a cereblon E3 ubiquitin ligase modulating agent (CELMoD), acts as a molecular glue. Mezigdomide shows high a ffinity to cereblon, resulting in potent antimyeloma activity[1]. 1 * 25 mg
  MDEXHY-129395
Frequently Bought Together