Order Entry
United Kingdom
ContactUsLinkComponent
proTAME, a cell-permeable proagent form of TAME, is an anaphase promoting complex/cyclosome (APC/C) inhibitor. proTAME causes cell c ycle arrest in metaphase[1][2]. 1 * 50 mg
  MDEXHY-124955
proTAME, a cell-permeable proagent form of TAME, is an anaphase promoting complex/cyclosome (APC/C) inhibitor. proTAME causes cell c ycle arrest in metaphase[1][2]. 1 * 50 mg
  MDEXHY-124955
Frequently Bought Together