Order Entry
United Kingdom
ContactUsLinkComponent
RMC-9805 (KRAS G12D inhibitor 18) is a KRAS G12D inhibitor. RMC-9805 is orally active. RMC-9805 inhibits RAS signaling to induce apo ptosis in KRAS G12D mutant cancer cells[1][2]. 1 * 10 mg
Catalog # MDEXHY-156819
Supplier:  MEDCHEMEXPRESS
RMC-9805 (KRAS G12D inhibitor 18) is a KRAS G12D inhibitor. RMC-9805 is orally active. RMC-9805 inhibits RAS signaling to induce apo ptosis in KRAS G12D mutant cancer cells[1][2]. 1 * 10 mg
Catalog # MDEXHY-156819
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together