Order Entry
United Kingdom
ContactUsLinkComponent
(R)-MG-132 ((S,R,S)-(-)-MG-132) is the enantiomer of MG-132. (R)-MG-132 is a proteasome inhibitor with weaker cell cytotoxicity than MG-132[1]. 1 * 100 mg
Catalog # MDEXHY-13259C
Supplier:  MEDCHEMEXPRESS
(R)-MG-132 ((S,R,S)-(-)-MG-132) is the enantiomer of MG-132. (R)-MG-132 is a proteasome inhibitor with weaker cell cytotoxicity than MG-132[1]. 1 * 100 mg
Catalog # MDEXHY-13259C
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together