Order Entry
United Kingdom
ContactUsLinkComponent
Buparlisib (BKM120 NVP-BKM120) is a pan-class I PI3K inhibitor, with IC50s of 52, 166, 116 and 262 nM for p110α, p110β, p110δ and p 110γ, respectively. 1 * 5 mg
  MDEXHY-70063
Buparlisib (BKM120 NVP-BKM120) is a pan-class I PI3K inhibitor, with IC50s of 52, 166, 116 and 262 nM for p110α, p110β, p110δ and p 110γ, respectively. 1 * 5 mg
  MDEXHY-70063
Frequently Bought Together