Order Entry
United Kingdom
ContactUsLinkComponent
BAY-876 is an orally active and selective glucose transporter 1 (GLUT1) inhibitor with an IC50 of 2 nM. BAY-876 is >130-fold more se lective for GLUT1 than GLUT2, GLUT3, and GLUT4. BAY-876 is also a potent blocker of glycolytic metabolism and ovarian 1 * 100 mg
Catalog # MDEXHY-100017
Supplier:  MEDCHEMEXPRESS
BAY-876 is an orally active and selective glucose transporter 1 (GLUT1) inhibitor with an IC50 of 2 nM. BAY-876 is >130-fold more se lective for GLUT1 than GLUT2, GLUT3, and GLUT4. BAY-876 is also a potent blocker of glycolytic metabolism and ovarian 1 * 100 mg
Catalog # MDEXHY-100017
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together