Order Entry
United Kingdom
ContactUsLinkComponent
Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Temsirolimus activates autophagy and prevents deterioration of cardiac function in animal model[8]. 1 * 10 mg
Catalog # MDEXHY-50910
Supplier:  MEDCHEMEXPRESS
Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Temsirolimus activates autophagy and prevents deterioration of cardiac function in animal model[8]. 1 * 10 mg
Catalog # MDEXHY-50910
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together