Order Entry
United Kingdom
ContactUsLinkComponent
EIPA (L593754) is an orally active TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA also enhances autophagy by inhibiting Na+/H +-exchanger 3 (NHE3). EIPA inhibits macropinocytosis as well. EIPA can be used in the research of inflammation and ca 1 * 100 mg
Catalog # MDEXHY-101840
Supplier:  MEDCHEMEXPRESS
EIPA (L593754) is an orally active TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA also enhances autophagy by inhibiting Na+/H +-exchanger 3 (NHE3). EIPA inhibits macropinocytosis as well. EIPA can be used in the research of inflammation and ca 1 * 100 mg
Catalog # MDEXHY-101840
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together