Order Entry
United Kingdom
ContactUsLinkComponent
Verapamil ((±)-Verapamil) is a calcium channel blocker and a potent and orally active first-generation P-glycoprotein (P-gp) inhibit or. Verapamil also inhibits CYP3A4. Verapamil has the potential for high blood pressure, heart arrhythmias and angina 1 * 50 mg
Catalog # MDEXHY-14275
Supplier:  MEDCHEMEXPRESS
Verapamil ((±)-Verapamil) is a calcium channel blocker and a potent and orally active first-generation P-glycoprotein (P-gp) inhibit or. Verapamil also inhibits CYP3A4. Verapamil has the potential for high blood pressure, heart arrhythmias and angina 1 * 50 mg
Catalog # MDEXHY-14275
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together