Order Entry
United Kingdom
ContactUsLinkComponent
Celecoxib,a selective non-steroidal anti-inflammatory drug (NSAID), is a selective COX-2 inhibitor with an IC50 of 40 nM. 1 * 100 mg
Catalog # MDEXHY-14398
Supplier:  MEDCHEMEXPRESS
Celecoxib,a selective non-steroidal anti-inflammatory drug (NSAID), is a selective COX-2 inhibitor with an IC50 of 40 nM. 1 * 100 mg
Catalog # MDEXHY-14398
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together