Order Entry
United Kingdom
ContactUsLinkComponent
740 Y-P (740YPDGFR PDGFR 740Y-P) is a potent and cell-permeable PI3K activator. 740 Y-P readily binds GST fusion proteins containin g both the N- and C- terminal SH2 domains of p85 but fails to bind GST alone[1]. 1 * 5 mg
Catalog # MDEXHY-P0175
Supplier:  MEDCHEMEXPRESS
740 Y-P (740YPDGFR PDGFR 740Y-P) is a potent and cell-permeable PI3K activator. 740 Y-P readily binds GST fusion proteins containin g both the N- and C- terminal SH2 domains of p85 but fails to bind GST alone[1]. 1 * 5 mg
Catalog # MDEXHY-P0175
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together