Order Entry
United Kingdom
ContactUsLinkComponent
ML-210 is a selective and covalent glutathione peroxidase 4 (GPX4) inhibitor with an EC50 of 30 nM. ML-210 binds the GPX4 selenocyst eine residue. ML-210 has anti-cancer activity[1][2]. 1 * 50 mg
Catalog # MDEXHY-100003
Supplier:  MEDCHEMEXPRESS
ML-210 is a selective and covalent glutathione peroxidase 4 (GPX4) inhibitor with an EC50 of 30 nM. ML-210 binds the GPX4 selenocyst eine residue. ML-210 has anti-cancer activity[1][2]. 1 * 50 mg
Catalog # MDEXHY-100003
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together