Order Entry
United Kingdom
ContactUsLinkComponent
TGF beta 1/TGFB1 Protein is initially identified as a growth factor that induces the growth of rodent fibroblasts. TGF beta 1/TGFB1 Protein inhibits the cell cycle in the G1 phase. TGF beta 1/TGFB1 is an endogenous factor controlling apoptosis in nor 1 * 1 Each
Catalog # MDEXHY-P7118
Supplier:  MEDCHEMEXPRESS
TGF beta 1/TGFB1 Protein is initially identified as a growth factor that induces the growth of rodent fibroblasts. TGF beta 1/TGFB1 Protein inhibits the cell cycle in the G1 phase. TGF beta 1/TGFB1 is an endogenous factor controlling apoptosis in nor 1 * 1 Each
Catalog # MDEXHY-P7118
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together