Order Entry
United Kingdom
ContactUsLinkComponent
Retatrutide (LY3437943) is a triple agonist peptide of the glucagon receptor (GCGR), glucosedependent insulinotropic polypeptide rec eptor (GIPR), and glucagon-like peptide-1 receptor (GLP-1R). Retatrutide binds human GCGR, GIPR, and GLP-1R with EC50 1 * 10 mg
Catalog # MDEXHY-P3506
Supplier:  MEDCHEMEXPRESS
Retatrutide (LY3437943) is a triple agonist peptide of the glucagon receptor (GCGR), glucosedependent insulinotropic polypeptide rec eptor (GIPR), and glucagon-like peptide-1 receptor (GLP-1R). Retatrutide binds human GCGR, GIPR, and GLP-1R with EC50 1 * 10 mg
Catalog # MDEXHY-P3506
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together