Order Entry
United Kingdom
ContactUsLinkComponent
Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor[1]. Doxycycline hyclate shows antibacterial activity and anti-cancer cell proliferation activity[1][2][3][4][5]. 1 * 100 mg
Catalog # MDEXHY-N0565B
Supplier:  MEDCHEMEXPRESS
Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor[1]. Doxycycline hyclate shows antibacterial activity and anti-cancer cell proliferation activity[1][2][3][4][5]. 1 * 100 mg
Catalog # MDEXHY-N0565B
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together