Order Entry
United Kingdom
ContactUsLinkComponent
Lorlatinib (PF-06463922) is a selective, orally active, brain-penetrant and ATP-competitive ROS1/ALK inhibitor with anticancer activ ity. Lorlatinib has Kis of L1196M, respectively. Lorlatinib targets to EML4-ALK, and inhibits ALK phosphorylation wit 1 * 10 mg
  MDEXHY-12215
Lorlatinib (PF-06463922) is a selective, orally active, brain-penetrant and ATP-competitive ROS1/ALK inhibitor with anticancer activ ity. Lorlatinib has Kis of L1196M, respectively. Lorlatinib targets to EML4-ALK, and inhibits ALK phosphorylation wit 1 * 10 mg
  MDEXHY-12215
Frequently Bought Together