Order Entry
United Kingdom
ContactUsLinkComponent
FGF-2 is a member of the fibroblast family involved in bone healing, cartilage repair, bone repair, and nerve regeneration. FGF-2 is also a mitotic promoter that accelerates cell proliferation. FGF-2 regulates immune processes by specifically target 1 * 1 Each
Catalog # MDEXHY-P7004
Supplier:  MEDCHEMEXPRESS
FGF-2 is a member of the fibroblast family involved in bone healing, cartilage repair, bone repair, and nerve regeneration. FGF-2 is also a mitotic promoter that accelerates cell proliferation. FGF-2 regulates immune processes by specifically target 1 * 1 Each
Catalog # MDEXHY-P7004
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together