Order Entry
United Kingdom
ContactUsLinkComponent
Entinostat is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3, re spectively. 1 * 5 mg
Catalog # MDEXHY-12163
Supplier:  MEDCHEMEXPRESS
Entinostat is an oral and selective class I HDAC inhibitor, with IC50s of 243 nM, 453 nM, and 248 nM for HDAC1, HDAC2, and HDAC3, re spectively. 1 * 5 mg
Catalog # MDEXHY-12163
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together