Order Entry
United Kingdom
ContactUsLinkComponent
GSK-872 is a RIPK3 inhibitor, which binds RIP3 kinase domain with an IC50 of 1.8 nM, and inhibits kinase activity with an IC50 of 1. 3 nM. GSK-872 decreases the RIPK3-mediated necroptosis and subsequent cytoplasmic translocation and expression of HMG 1 * 1 mg
Catalog # MDEXHY-101872
Supplier:  MEDCHEMEXPRESS
GSK-872 is a RIPK3 inhibitor, which binds RIP3 kinase domain with an IC50 of 1.8 nM, and inhibits kinase activity with an IC50 of 1. 3 nM. GSK-872 decreases the RIPK3-mediated necroptosis and subsequent cytoplasmic translocation and expression of HMG 1 * 1 mg
Catalog # MDEXHY-101872
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together