Order Entry
United Kingdom
ContactUsLinkComponent
AICAR (Acadesine) is an adenosine analog and a AMPK activator. AICAR regulates the glucose and lipid metabolism, and inhibits proinf lammatory cytokines and iNOS production. AICAR is also an autophagy, YAP and mitophagy inhibitor[1][2]. 1 * 50 mg
Catalog # MDEXHY-13417
Supplier:  MEDCHEMEXPRESS
AICAR (Acadesine) is an adenosine analog and a AMPK activator. AICAR regulates the glucose and lipid metabolism, and inhibits proinf lammatory cytokines and iNOS production. AICAR is also an autophagy, YAP and mitophagy inhibitor[1][2]. 1 * 50 mg
Catalog # MDEXHY-13417
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together