Order Entry
United Kingdom
ContactUsLinkComponent
Volasertib (BI 6727) is an orally active, highly potent and ATP-competitive Polo-like kinase 1 (PLK1) inhibitor with an IC50 of 0.87 nM. Volasertib inhibits PLK2 and PLK3 with IC50s of 5 and 56 nM, respectively. Volasertib induces mitotic arrest and 1 * 5 mg
Catalog # MDEXHY-12137
Supplier:  MEDCHEMEXPRESS
Volasertib (BI 6727) is an orally active, highly potent and ATP-competitive Polo-like kinase 1 (PLK1) inhibitor with an IC50 of 0.87 nM. Volasertib inhibits PLK2 and PLK3 with IC50s of 5 and 56 nM, respectively. Volasertib induces mitotic arrest and 1 * 5 mg
Catalog # MDEXHY-12137
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together