Order Entry
United Kingdom
ContactUsLinkComponent
Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'. 1 * 100 mg
Catalog # MDEXHY-50855
Supplier:  MEDCHEMEXPRESS
Silmitasertib (CX-4945) is an orally bioavailable, highly selective and potent CK2 inhibitor, with IC50 values of 1 nM against CK2α and CK2α'. 1 * 100 mg
Catalog # MDEXHY-50855
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together