Order Entry
United Kingdom
ContactUsLinkComponent
Baricitinib (LY3009104 INCB028050) is a selective and orally bioavailable JAK1 and JAK2 inhibitor with IC50s of 5.9 nM and 5.7 nM, respectively. 1 * 10 mg
  MDEXHY-15315
Baricitinib (LY3009104 INCB028050) is a selective and orally bioavailable JAK1 and JAK2 inhibitor with IC50s of 5.9 nM and 5.7 nM, respectively. 1 * 10 mg
  MDEXHY-15315
Frequently Bought Together