Order Entry
United Kingdom
ContactUsLinkComponent
Disulfiram (Tetraethylthiuram disulfide) is a specific inhibitor of aldehyde-dehydrogenase (ALDH1), used for the treatment of chroni c alcoholism by producing an acute sensitivity to alcohol. Disulfiram inhibits gasdermin D (GSDMD) pore formation in 1 * 5 g
Catalog # MDEXHY-B0240
Supplier:  MEDCHEMEXPRESS
Disulfiram (Tetraethylthiuram disulfide) is a specific inhibitor of aldehyde-dehydrogenase (ALDH1), used for the treatment of chroni c alcoholism by producing an acute sensitivity to alcohol. Disulfiram inhibits gasdermin D (GSDMD) pore formation in 1 * 5 g
Catalog # MDEXHY-B0240
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together