Order Entry
United Kingdom
ContactUsLinkComponent
Valproic acid (VPA) is an orally active HDAC inhibitor, with IC50 in the range of 0.5 and 2 mM. Valproic acid inhibits HDAC1 (IC50, 400 μM), and induces proteasomal degradation of HDAC2. Valproic acid activates Notch1 signaling and inhibits prolifera 1 * 25 g
Catalog # MDEXHY-10585
Supplier:  MEDCHEMEXPRESS
Valproic acid (VPA) is an orally active HDAC inhibitor, with IC50 in the range of 0.5 and 2 mM. Valproic acid inhibits HDAC1 (IC50, 400 μM), and induces proteasomal degradation of HDAC2. Valproic acid activates Notch1 signaling and inhibits prolifera 1 * 25 g
Catalog # MDEXHY-10585
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together