Order Entry
United Kingdom
ContactUsLinkComponent
Hydroxyurea is a cell apoptosis inducer that inhibit DNA synthesis through inhibition of ribonucleotide reductase. Hydroxyurea shows anti-orthopoxvirus activity. 1 * 500 mg
Catalog # MDEXHY-B0313
Supplier:  MEDCHEMEXPRESS
Hydroxyurea is a cell apoptosis inducer that inhibit DNA synthesis through inhibition of ribonucleotide reductase. Hydroxyurea shows anti-orthopoxvirus activity. 1 * 500 mg
Catalog # MDEXHY-B0313
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together