Order Entry
United Kingdom
ContactUsLinkComponent
Z-VAD(OMe)-FMK (Z-Val-Ala-Asp(OMe)-FMK) is a cell-permeable and irreversible pan-caspase inhibitor[1]. Z-VAD(OMe)-FMK is an ubiquiti n carboxy-terminal hydrolase L1 (UCHL1) inhibitor. Z-VAD(OMe)-FMK irreversibly modifies UCHL1 by targeting the active 1 * 50 mg
Catalog # MDEXHY-16658
Supplier:  MEDCHEMEXPRESS
Z-VAD(OMe)-FMK (Z-Val-Ala-Asp(OMe)-FMK) is a cell-permeable and irreversible pan-caspase inhibitor[1]. Z-VAD(OMe)-FMK is an ubiquiti n carboxy-terminal hydrolase L1 (UCHL1) inhibitor. Z-VAD(OMe)-FMK irreversibly modifies UCHL1 by targeting the active 1 * 50 mg
Catalog # MDEXHY-16658
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together