Order Entry
United Kingdom
ContactUsLinkComponent
Nirmatrelvir (PF-07321332) is a potent and orally active SARS-CoV 3C-like protease (3CLPRO) inhibitor. Nirmatrelvir (PF-07321332) ta rgets to the SARS-CoV-2 virus and can be used for COVID-19 research[1]. 1 * 50 mg
Catalog # MDEXHY-138687
Supplier:  MEDCHEMEXPRESS
Nirmatrelvir (PF-07321332) is a potent and orally active SARS-CoV 3C-like protease (3CLPRO) inhibitor. Nirmatrelvir (PF-07321332) ta rgets to the SARS-CoV-2 virus and can be used for COVID-19 research[1]. 1 * 50 mg
Catalog # MDEXHY-138687
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together